Order Entry
Italy
ContactUsLinkComponent
Ammonium bismuth citrate ≥99%, Reagent Grade
Toxic
Ammonium bismuth citrate ≥99%, Reagent Grade
Catalog # 0808-100G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Ammonium bismuth citrate ≥99%, Reagent Grade
Catalog # 0808-100G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Chloride Test:
    Pass
  • Copper Test:
    Pass
  • Lead:
    <= 5 ppm
  • Nitrate Test:
    Pass
  • Purity (as Bi, NH4+ & Citrate):
    >= 99%
  • Solubility (0.5%, Water):
    Pass
  • Sulfate Test:
    Pass

Specification Test Results

About this item

 :
 :433.12 g/mol
EINECS:250-856-6
 :GHS06
Storage Temperature:Ambient
CAS Number:31886-41-6
Formula:C₆H₁₁BiNO₇
MDL Number:MFCD00036420