Order Entry
Italy
ContactUsLinkComponent
4,6-Diaminopyrimidine-2-thiol for synthesis, Sigma-Aldrich®
Irritant
4,6-Diaminopyrimidine-2-thiol for synthesis, Sigma-Aldrich®
Catalog # 8.14037.0025
Supplier:  Merck
CAS Number:  
Irritant
4,6-Diaminopyrimidine-2-thiol for synthesis, Sigma-Aldrich®
Catalog # 8.14037.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    280 °C
  • Molar Mass:
    142.17 g/mol

Specification Test Results

About this item

 :
 :142.18 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:213-721-2
 :GHS07
Storage Temperature:Ambient
CAS Number:1004-39-3
Formula:C₄H₆N₄S
MDL Number:MFCD00006078