Order Entry
Italy
ContactUsLinkComponent
(Ethoxycarbonylmethyl)triphenylphosphonium bromide 98%
Irritant
(Ethoxycarbonylmethyl)triphenylphosphonium bromide 98%
Catalog # ACRO108300250
CAS Number:  
undefined
Irritant
(Ethoxycarbonylmethyl)triphenylphosphonium bromide 98%
Catalog # ACRO108300250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥97.5 %
  • GC:
    ≥98.5 %

Specification Test Results

About this item

 :
 :429.30 g/mol
Melting Pt:158 °C (dec.)
EINECS:216-230-1
 :GHS07
Storage Temperature:Ambient
CAS Number:1530-45-6
Formula:C₂₂H₂₂BrO₂P
MDL Number:MFCD00011835