
Bacitracin, potency min. 60 Units/mg
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1405-87-4
:
Formula:C₆₆H₁₀₃N₁₇O₁₆S₁
MDL Number:MFCD00062640
:1421.60 g/mol
Melting Pt:221 °C…225 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white to beige Powder
- Infrared spectrum:Conforms
- Loss on drying:≤5.0 % (1 g, 60°C, 3 h) (P2O5) (not exceeding 0.1 kPa)
- Sulfated ash:≤1.0 %
- Potency:≥60 IU/mg (on dried substance)
- Related substances:(Eur. Pharm. (HPLC)) Passes test for composition
Specification Test Results
Frequently Bought Together