Spermine tetrahydrochloride, Millipore®
Catalog # 5677-5
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:348.19 g/mol
EINECS:206-189-8
CAS Number:306-67-2
Formula:C₁₀H₃₀Cl₄N₄
MDL Number:MFCD00012914