Order Entry
Italy
ContactUsLinkComponent
2,6-Dibromoanthracene ≥98.0% (by HPLC)
2,6-Dibromoanthracene ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
undefined
2,6-Dibromoanthracene ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%

Specification Test Results

Related Information

Frequently Bought Together