Order Entry
Italy
ContactUsLinkComponent
 
Organic reference standard, Benz[a]anthracene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERL20545000CY
 :  
Organic reference standard, Benz[a]anthracene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERL20545000CY
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    56-55-3
  • Description:
    Benz[a]anthracene 10 µg/ml in cyclohexane
  • Formula:
    C₁₈H₁₂
  • Description:
    Benz[a]anthracene