Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
α-D-Glucose
α-D-Glucose
α-D-Glucose
α-D-Glucose
EHERC14027000
:
EHRENSTORFER
:
α-D-Glucose
EHERC14027000
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,25 g
Seq:
3029
:
:
180.1559 g/mol
:
207-757-8
:
492-62-6
:
C₆H₁₂O₆
:
MFCD00063774