2-Deoxy-D-glucose 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:154-17-6
:MFCD00007013
Formula:C₆H₁₂O₅
MDL Number:MFCD00151328
:164.16 g/mol
Melting Pt:146…147 °C
Storage Temperature:Ambient
EINECS:205-823-0
Specification Test Results
- Appearance:White to cream Crystalline powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤1 %
- Solubility:(2 % in water) Clear colorless to light yellow
- TLC:≥98.5 %
- Residue after ignition:≤1 %
- Specific optical rotation:+44.0° to +48.0° (20°C, 589 nm) (c=1, H2O) (after 24 h)
- Microbial contamination:Passes test
- pH:3.0 to 5.0 (2 % in water)
Specification Test Results
Frequently Bought Together