Hydroxypropyl cellulose, M.W. 100000, 75 - 150 mPa.s (5 % in H₂O)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9004-64-2
:
MDL Number:MFCD00132688
Merck Index:14,4841
Storage Temperature:Ambient
Related Information
Frequently Bought Together