Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cyclopentylacetic acid
Cyclopentylacetic acid for synthesis
Cyclopentylacetic acid for synthesis
Cyclopentylacetic acid for synthesis
8.41330.0010
:
Merck
:
Cyclopentylacetic acid for synthesis
8.41330.0010
:
Merck
:
Pack type:
Glass bottle
Pk:
10 ml
Seq:
2275
:
:
128.17 g/mol
:
214-368-7
:
1123-00-8
:
C₇H₁₂O₂
:
MFCD00001387