Order Entry
Italy
ContactUsLinkComponent
Cetrimonium chloride 50% in 2-propanol 60% (v/v)/water 40% (v/v) for synthesis, Sigma-Aldrich®
Flammable
Corrosive
Irritant
Environment
Cetrimonium chloride 50% in 2-propanol 60% (v/v)/water 40% (v/v) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Corrosive
Irritant
Environment
Cetrimonium chloride 50% in 2-propanol 60% (v/v)/water 40% (v/v) for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Boiling Point:
    80 - 90 °C
  • Flash Point:
    15 °C
  • Refractive Index:
    1.3778 (20 °C, 589 nm)
  • Density:
    0.89 g/cm3 (20 °C)
  • pH:
    5 - 7 (10 g/l, H2O, 20 °C)
  • Synonyms:
    Cetyltrimethylammonium chloride solution, Trimethylhexadecylammonium chloride

Specification Test Results

Related Information

Frequently Bought Together