Order Entry
Italy
ContactUsLinkComponent
Sodium bisulphite, Reagent Grade
Sodium bisulphite, Reagent Grade
Catalog # N822-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium bisulphite, Reagent Grade
Catalog # N822-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Chloride Content:
    <0.02%
  • Heavy Metals (as Pb):
    <0.001%
  • Insoluble Matter:
    <0.005%
  • Iron:
    <0.002%
  • Titration by Na₂SO₃ (%SO₂):
    >58.5%

Specification Test Results

About this item

 :
Merck Index:12,08731
Melting Pt:300 °C
Density:2.476 g/cm³ (20 °C)
ADR:8,III
 :104.06 g/mol
EINECS:231-548-0
UN:2693
 :GHS07
 :GHS05
Storage Temperature:Ambient
CAS Number:7631-90-5
Formula:HNaO₃S
MDL Number:MFCD00167602