Order Entry
Italy
ContactUsLinkComponent
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt)
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt)
Catalog # 10016.
Supplier:  Biotium
CAS Number:  
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt)
Catalog # 10016.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :521.79 g/mol
Melting Pt:230 °C
Storage Temperature:Freezer
CAS Number:114162-64-0
Formula:C₂₀H₂₆BrClN₂O₇
MDL Number:MFCD00058543