Order Entry
Italy
ContactUsLinkComponent
Cerium standard solution, 1,000 mg/L Ce in dil. nitric acid (from Ce(NO₃)₃.6H₂O) standard for AAS and ICP
Cerium standard solution, 1,000 mg/L Ce in dil. nitric acid (from Ce(NO₃)₃.6H₂O) standard for AAS and ICP
  PERCN9300110
 :  
Cerium standard solution, 1,000 mg/L Ce in dil. nitric acid (from Ce(NO₃)₃.6H₂O) standard for AAS and ICP
  PERCN9300110
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Cerium
  • Symbol:
    Ce