Order Entry
Italy
Orders LinkContactUsLinkComponent
N-Hydroxysuccinimide

N-Hydroxysuccinimide

N-Hydroxysuccinimide
Certificates
N-Hydroxysuccinimide

2 Options Available Below

Product Details & Documents
Synonyms:  1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu
  • CAS Number:6066-82-6
  • Formula:C₄H₅NO₃
  • MDL Number:MFCD00005516
  • Molecular Weight:115.09 g/mol
  • Boiling Pt:363 °C (1013 hPa)
  • Melting Pt:93…95 °C
  • Storage Temperature:Refrigerator
  • EINECS:228-001-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 28,60 €
    28,60 €
    1 * 25 g
       28,60 €

  • Plastic bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 84,50 €
    84,50 €
    1 * 100 g
       84,50 €