Safety canisters with screw cap
Supplier: DUPERTHAL
Certificates
About this item
Safety canister for transport of hazardous substances, made of 1.4571 polished steel with teflon seal.
- Tight closure, captive cap on flexible shaft
- Flame arrester in can neck
- Can be used in-house with measuring tap
- 38 mm pipe thread
Markings according to GetStoffV and BGV A8, additional markings according to ADR/RID.
Frequently Bought Together