Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) 90%
Catalog # ACRO226310010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤15 % (105°C, 5 hrs)
- HPLC:≥88 %
- UV Lambda max:262 nm (+/- 2 nm)
- pH:3.0 to 7.0 (0.5 % in water)
- Absorbance scale:0.33 to 0.43 (A280/A260)
- Chloride (Cl):≤2000 ppm
- Heavy metals (as Pb):≤20 ppm
Specification Test Results
About this item
:
:550.09 g/mol
Melting Pt:>140 °C (dec.)
Flash Pt:>110 °C
Storage Temperature:Freezer
EINECS:243-347-5
CAS Number:19817-92-6
Formula:C₉H₁₂N₂Na₃O₁₅P₃
MDL Number:MFCD00150714