Rain jacket, Bielefeld 4265
Supplier: SIOEN
About this item
Waterproof and windproof jacket made of Flexothane® Essential (100% polyester knitting with PU coating). With high frequency welded seams and rain gutter.
- Straight collar, foldaway hood in collar, zip closure under flap with press studs
- Two inset pockets
- Raglan sleeves, sleeve narrowing by press studs
- Back ventilation
Weight: 170 g/m²
: ENV 343 (3-1)
18 Options Available Below
Product Details & Documents
Waterproof and windproof jacket made of Flexothane® Essential (100% polyester knitting with PU coating). With high frequency welded seams and rain gutter.
- Straight collar, foldaway hood in collar, zip closure under flap with press studs
- Two inset pockets
- Raglan sleeves, sleeve narrowing by press studs
- Back ventilation
Weight: 170 g/m²
: ENV 343 (3-1)
Specifications for Product Family
Specifications
- Catalog No:
- 113-5931
- 113-5930
- 113-5929
- 113-5933
- 113-5934
- 113-5935
- 113-07122
- 113-07121
- 113-07120
- 113-07125
- 113-07126
- 113-07127
- 113-07124
- 113-07123
- 113-5932
- 113-5936
- 113-5937
- 113-5938
- Model:
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- 4265 Bielefeld
- Garment application:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Gender:
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- 100% polyester/PU coating
- Weight:
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
Specifications
Product Family Options
Product Information
- ColourClothing SizeAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.113-5931Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-5930Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-5929Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-5933Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-5934Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-5935Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-07122Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-07121Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-07120Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-07125Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-07126Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-07127Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-07124Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-07123Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-5932Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-5936Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-5937Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m² - Specifications:
More
Specifications
Cat. No.113-5938Model4265 BielefeldGarment applicationOutdoorRisk areaRain (waterproof)GenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric100% polyester/PU coatingWeight170 g/m²



