These testo thermo-hygrometers are convenient and easy-to operate pocket-sized material moisture measuring instrument. It is used for the measurement of wood moisture, characteristic curves are available for beech, spruce, larch, oak, pine and maple. For locating wet spots in building materials, curves are additionally stored for cement screed, plaster, anhydrite screed, cement mortar, lime mortar and bricks.
- Precise measurement of wood moisture
- Characteristics curves for locating wet spots in building materials
- Hold-function for convenient read-out of measurement values
- Display illumination
The testo 606-2 model measures air humidity and temperature in addition to material moisture. This enables drying conditions to be determined quickly and reliably.
The testo 606-1 is a compact moisture meter that measures the moisture content in wood, walls and other surfaces and materials and fits snugly in any pocket. Ideal for both professionals and private users who appreciate the precision offered by such a professional tool.
- Catalog No:
- 620-3309
- 620-3310
- Batteries:
- 2 AAA micro batteries
- 2 AAA micro batteries
- W×D×H:
- 119×46×25 mm
- 119×46×25 mm
- Weight:
- 90 g
- 90 g