Order Entry
Italy
ContactUsLinkComponent
 
Irritant
Ampicillin trihydrate
Catalog # EHERC10243080
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
Ampicillin trihydrate
Catalog # EHERC10243080
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :403.45 g/mol
EINECS:200-709-7
 :GHS07
Storage Temperature:Refrigerator
CAS Number:7177-48-2
Formula:C₁₆H₁₉N₃O₄S·3H₂O
MDL Number:MFCD00072036