Order Entry
Italy
ContactUsLinkComponent
 
Flammable
Corrosive
Health
Irritant
Tetrabutylammonium dihydrogentrifluoride 50-55% (w/w) in 1,2-dichloroethane
Catalog # ACRO268780025
CAS Number:  
undefined
Flammable
Corrosive
Health
Irritant
Tetrabutylammonium dihydrogentrifluoride 50-55% (w/w) in 1,2-dichloroethane
Catalog # ACRO268780025
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 g
  • Seq:
    3122

Specifications

Specification Test Results

  • Appearance:
    Clear light yellow to brown Solution
  • Infrared spectrum:
    Conforms
  • NMR:
    Consistent with structure
  • Strength:
    50 to 55 % (NMR)

Specification Test Results

About this item

 :
 :301.48 g/mol
Flash Pt:13 °C
Storage Temperature:Refrigerator
UN:2924
ADR:3,II
 :GHS02
 :GHS05
 :GHS08
 :GHS07
CAS Number:99337-56-1
Formula:C₁₆H₃₈F₃N
MDL Number:MFCD00145365