S-Methylisothiouronium sulphate
Catalog # CAYM81300-1
Supplier: Cayman Chemical
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:278.38 g/mol
EINECS:212-759-7
CAS Number:867-44-7
Formula:C₄H₁₂N₄S₂·H₂SO₄
MDL Number:MFCD00013055