Order Entry
Italy
ContactUsLinkComponent
Herbicide reference standard, Hexazinone 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Herbicide reference standard, Hexazinone 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL14200000AL
Supplier:  EHRENSTORFER
CAS Number:  
Herbicide reference standard, Hexazinone 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL14200000AL
Supplier:  EHRENSTORFER
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    51235-04-2
  • Description:
    Hexazinone 10 µg/ml in acetonitrile
  • Formula:
    C₁₂H₂₀N₄O₂
  • Description:
    Hexazinone

Specifications

About this item