DL-Menthol 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: rel-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol
- CAS Number:89-78-1
- Formula:C₁₀H₂₀O
- MDL Number:MFCD00001484
- Molecular Weight:156.27 g/mol
- Melting Pt:32…36 °C
- Density:0.89 g/cm³
- Flash Pt:92 °C
- Storage Temperature:Ambient
- EINECS:201-939-0
3 Options Available Below
Product Details & Documents
Synonyms: rel-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol
- CAS Number:89-78-1
- Formula:C₁₀H₂₀O
- MDL Number:MFCD00001484
- Molecular Weight:156.27 g/mol
- Melting Pt:32…36 °C
- Density:0.89 g/cm³
- Flash Pt:92 °C
- Storage Temperature:Ambient
- EINECS:201-939-0
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Colorless to white Crystalline solid
- Infrared spectrum:Conforms
- Melting point:32 to 36 °C
- GC:≥98.5 %
- Specific optical rotation:-2° to +2° (20 °C, 589 nm) (c=10, EtOH)
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
- Cadmium (Cd):≤0.05 ppm
- Calcium (Ca):≤0.5 ppm
- Chromium (Cr):≤0.02 ppm
- Cobalt (Co):≤0.02 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Manganese (Mn):≤0.02 ppm
- Nickel (Ni):≤0.02 ppm
- Zinc (Zn):≤0.1 ppm
- Acidity (CH3COOH):≤0.01 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



