Order Entry
Italy
ContactUsLinkComponent
Vitamin K1 (phytomenadione, phylloquinone) 1% lactose trituration
Vitamin K1 (phytomenadione, phylloquinone) 1% lactose trituration
  5.01758.5000
 :  Merck
 :  
Vitamin K1 (phytomenadione, phylloquinone) 1% lactose trituration
  5.01758.5000
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 kg
  • Seq:
    4114

 

 

 :
 :450.6958 g/mol
 :201-564-2
 :84-80-0
 :81818-54-4
 :C₃₁H₄₆O₂
 :MFCD00214063