Order Entry
Italy
ContactUsLinkComponent
γ-Cyclodextrine hydrate 99%
γ-Cyclodextrine hydrate 99%
Certificates
undefined
γ-Cyclodextrine hydrate 99%
Specification Test Results
  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤11 %
  • HPLC:
    ≥98.5 % (ELSD)