Order Entry
Italy
ContactUsLinkComponent
2-(Allylthio)nicotinic acid ≥98%
Irritant
2-(Allylthio)nicotinic acid ≥98%
Catalog # L07090.06
CAS Number:  
Irritant
2-(Allylthio)nicotinic acid ≥98%
Catalog # L07090.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

CAS Number:175135-25-8
 :
Formula:C₉H₉NO₂S
 :195.24 g/mol
Melting Pt:144…149 °C
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD00051681