1,8-Bis(dimethylamino)naphthalene ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 20734-58-1 Molecular Formula: C14H18N2 Formula Weight: 214.31 Flash Point: 110°(230°F) Melting Point: 47-51° Boiling Point: 144-145°/7mm Beilstein Reference: 396782 UN No.: 3263 MDL No.: MFCD00003920
Synonyms: N1,N1,N8,N8-Tetramethylnaphthalene-1,8-diamine
- CAS Number:20734-58-1
- Formula:C₁₄H₁₈N₂
- MDL Number:MFCD00003920
- Molecular Weight:214.31 g/mol
- Boiling Pt:144…145 °C (7 mmHg)
- Melting Pt:47…51 °C
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:244-001-6
- UN:3263
- ADR:8,III
2 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 20734-58-1 Molecular Formula: C14H18N2 Formula Weight: 214.31 Flash Point: 110°(230°F) Melting Point: 47-51° Boiling Point: 144-145°/7mm Beilstein Reference: 396782 UN No.: 3263 MDL No.: MFCD00003920
Synonyms: N1,N1,N8,N8-Tetramethylnaphthalene-1,8-diamine
- CAS Number:20734-58-1
- Formula:C₁₄H₁₈N₂
- MDL Number:MFCD00003920
- Molecular Weight:214.31 g/mol
- Boiling Pt:144…145 °C (7 mmHg)
- Melting Pt:47…51 °C
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:244-001-6
- UN:3263
- ADR:8,III



