

1,8-Bis(dimethylamino)naphthalene 99%
:20734-58-1
:
:C₁₄H₁₈N₂
:MFCD00003920
:214.31 g/mol
:144…145 °C (5hPa)
:46…50 °C
:>110 °C
:Ambient
:244-001-6
:3259
:8,III
:GHS07
:GHS05
- Appearance:Light yellow to beige to pink or brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:46 to 50 °C
- GC:≥98.5 %
Frequently Bought Together