Inserisci ordine
Italy
ContactUsLinkComponent
2-Methoxy-5-(trifluoromethyl)benzaldehyde ≥98%
Irritant
2-Methoxy-5-(trifluoromethyl)benzaldehyde ≥98%
Catalog # H26135.03
CAS Number:  
Irritant
2-Methoxy-5-(trifluoromethyl)benzaldehyde ≥98%
Catalog # H26135.03
CAS Number:  

Specifications

  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:146539-83-5
 :
Formula:C₉H₇F₃O₂
 :204.15 g/mol
Melting Pt:85…89 °C
EINECS:None
 :GHS07
Storage Temperature:Ambiente
MDL Number:MFCD03788528