4-Bromophenyl phenyl ether (BDE No. 3) ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:101-55-3
:
Formula:C₁₂H₉BrO
MDL Number:MFCD00000094
Boiling Pt:304…305 °C
Melting Pt:17…18 °C
Density:1.420
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:202-952-4
ADR:9,III
:249.11 g/mol
UN:3082
Related Information
Frequently Bought Together