Order Entry
Italy
ContactUsLinkComponent
2,6-Dichloroquinone-4-chloroimide, Sigma-Aldrich®
Flammable
Irritant
2,6-Dichloroquinone-4-chloroimide, Sigma-Aldrich®
Certificates
undefined
Flammable
Irritant
2,6-Dichloroquinone-4-chloroimide, Sigma-Aldrich®
Certificates

Specification Test Results

  • Appearance (Colour):
    Yellow to Orange
  • Appearance (Form):
    Powder or Crystals
  • Solubility (Turbidity):
    Clear
  • Solubility (Colour), 50 mg/ml, Ethanol:
    Yellow
  • Carbon:
    32.4 - 36.0%
  • Nitrogen:
    6.3 - 7.3%
  • Purity (TLC):
    ≥95 %
  • Suitability; Suitable For Use In The Detection Of Phenols:
    Suitable

Specification Test Results

Related Information

Frequently Bought Together