Tubing, perbunan rubber
Supplier: DEUTSCH NEUMANN
Certificates
About this item
Acrylonitrile-Butadiene (NBR), black.
- Hardness: 70 Shore A
- Temperatur resistance: −30 to +80 °C
- No inlay
- Resistant against oil.
- Dimension tolerances according to DIN ISO 3302-1 E2
Frequently Bought Together