Tubing, silicone, vacuum
Supplier: DEUTSCH NEUMANN
Certificates
About this item
Made of Silicone, translucent
- Very good chemical resistance
- Temperature range −60 to +200 °C
- Flexible
Certifications: Meet standards BFR XV, FDA CFR 177,2600, Europ.Pharmacopoeia
Frequently Bought Together