Laboratory glass cleaner, borer 24 LIQ
Liquid, highly alkaline cleaning concentrate for automated cleaning of laboratory glassware und reusable laboratory material.
- Rinsable without residues and guarantees therefore consistent and error-free results
- With a broad cleaning spectrum
- Perfect results even at low concentrations
- High cleaning performance due to excellent dispersing capabilities
- Contains active chlorine as a cleaning booster
- EDTA-, NTA- and tensidfree, no release of surfactants into waste water
- Rinsing with an acidic deconex® cleaner is recommended as the completion of the cleaning process
Frequently Bought Together