Order Entry
Italy
ContactUsLinkComponent
Lithium standard solution, concentrate, 1000 mg Li in water (from LiCl) Titrisol® standard for AAS, Supelco®
Lithium standard solution, concentrate, 1000 mg Li in water (from LiCl) Titrisol® standard for AAS, Supelco®
Catalog # 1.09934.0001
Supplier:  Merck
CAS Number:  
Lithium standard solution, concentrate, 1000 mg Li in water (from LiCl) Titrisol® standard for AAS, Supelco®
Catalog # 1.09934.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg
  • Matrix:
    H₂O
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • M-Clarity quality level:
    MQ100
  • Element:
    Lithium
  • Symbol:
    Li

Specifications

About this item

Certifications: Traceable to SRM from NIST