Order Entry
Italy
ContactUsLinkComponent
(R)-(+)-1,1'-Bi-2-naphthol for synthesis, Sigma-Aldrich®
Toxic
(R)-(+)-1,1'-Bi-2-naphthol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
(R)-(+)-1,1'-Bi-2-naphthol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    204 - 208 °C
  • Molar Mass:
    286.33 g/mol

Specification Test Results

Related Information

Frequently Bought Together