Pesticide reference standard, Captafol (1,2,3,6-Tetrahydro-N-(1,1,2,2-tetrachloroethylthio)phthalimide) 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA10950000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:2425-06-1
- Description:Captafol (1,2,3,6-Tetrahydro-N-(1,1,2,2-tetrachloroethylthio)phthalimide) 100 µg/ml in cyclohexane
- Formula:C₁₀H₉Cl₄NO₂S
- Description:Captafol (1,2,3,6-Tetrahydro-N-(1,1,2,2-tetrachloroethylthio)phthalimide)