

Diisopropyl azodicarboxylate ≥94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2446-83-5
:
Formula:C₈H₁₄N₂O₄
MDL Number:MFCD00008875
:202.21 g/mol
Boiling Pt:74…75 °C (0.3 mmHg)
Melting Pt:3…5 °C
Density:1.027
Flash Pt:160 °C (320 °F)
EINECS:219-502-8
ADR:9,III
:GHS07
:GHS09
Related Information
Frequently Bought Together