Order Entry
Italy
ContactUsLinkComponent
Di-tert-butyl azodicarboxylate 97%
Irritant
Flammable
Di-tert-butyl azodicarboxylate 97%
Certificates
undefined
Irritant
Flammable
Di-tert-butyl azodicarboxylate 97%

Specification Test Results

  • Appearance:
    Yellow Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    89 to 93 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together