Order Entry
Italy
ContactUsLinkComponent
 
Irritant
2-Chloro-N,N-dimethylacetamide for synthesis, Sigma-Aldrich®
Catalog # 8.20290.0025
Supplier:  Merck
CAS Number:  
Irritant
2-Chloro-N,N-dimethylacetamide for synthesis, Sigma-Aldrich®
Catalog # 8.20290.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 ml
  • Seq:
    2315

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Molar Mass:
    121.56 g/mol
  • Boiling Point:
    98 - 100 °C (15 hPa)
  • Density:
    1.18 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :121.57 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:220-224-4
 :GHS07
Storage Temperature:Ambient
CAS Number:2675-89-0
Formula:C₄H₈ClNO
MDL Number:MFCD00039360