


Hexafluoroacetone sesquihydrate 98%
Catalog # ACRO269970250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Clear light yellow to amber Liquid
- Infrared spectrum:Conforms
- Assay:≥97.5 %
Specification Test Results
About this item
:
:193.05 g/mol
Melting Pt:18…21 °C
Storage Temperature:Ambient
EINECS:211-676-3
UN:2552
ADR:6.1,II
:GHS05
:GHS06
:GHS08
CAS Number:13098-39-0
Formula:C₃F₆O·1.5H₂O
MDL Number:MFCD00149063