Herbicide reference standard, Propachlor 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA16380000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:1918-16-7
- Description:Propachlor 100 µg/ml in cyclohexane
- Environmentally Preferable:
- Formula:C₁₁H₁₄ClNO
- Description:Propachlor