Order Entry
Italy
ContactUsLinkComponent
Pesticide reference standard, Parathion-methyl (O,O-Diethyl O-4-nitrophenyl phosphorothioate) 100 µg/ml in toluene, Dr Ehrenstorfer
Pesticide reference standard, Parathion-methyl (O,O-Diethyl O-4-nitrophenyl phosphorothioate) 100 µg/ml in toluene, Dr Ehrenstorfer
Catalog # EHERXA15890000TO
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Parathion-methyl (O,O-Diethyl O-4-nitrophenyl phosphorothioate) 100 µg/ml in toluene, Dr Ehrenstorfer
Catalog # EHERXA15890000TO
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    298-00-0
  • Description:
    Parathion-methyl (O,O-Diethyl O-4-nitrophenyl phosphorothioate) 100 µg/ml in toluene
  • Formula:
    C₈H₁₀NO₅PS
  • Description:
    Parathion-methyl (O,O-Diethyl O-4-nitrophenyl phosphorothioate)

Specifications

About this item