Pesticide reference standard, Parathion-methyl (O,O-Diethyl O-4-nitrophenyl phosphorothioate) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Codice catalogo EHERXA15890000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:298-00-0
- Description:Parathion-methyl (O,O-Diethyl O-4-nitrophenyl phosphorothioate) 100 µg/ml in acetonitrile
- Formula:C₈H₁₀NO₅PS
- Description:Parathion-methyl (O,O-Diethyl O-4-nitrophenyl phosphorothioate)