

Tetrabutylammonium dihydrogentrifluoride tech. 90%, Technical Grade
Catalog # H27266.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
CAS Number:99337-56-1
:
Formula:C₁₆H₃₈F₃N
MDL Number:MFCD00145365
:301.48 g/mol
UN:3265
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Ambient