Order Entry
Italy
ContactUsLinkComponent
(2,4-Difluorophenyl)acetone 98%
Irritant
(2,4-Difluorophenyl)acetone 98%
Certificates
undefined
Irritant
(2,4-Difluorophenyl)acetone 98%

Specification Test Results

  • Appearance:
    Clear yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4770 to 1.4790 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together