2,2-Dichlorobutyrophenone for synthesis
Catalog # 8.14490.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 ml
- Seq:2210
Specifications
Specification Test Results
- Molar Mass:217.09 g/mol
- Boiling Point:132 - 136 °C
- Density:1.23 g/cm3 (20 °C)
- Synonyms:2,2-Dichloropropyl phenyl ketone
- Hill Formula:C10H10Cl2O
- Chemical Formula:C6H5COC(Cl2)CH2CH3
Specification Test Results
About this item
:
:M-Clarity™ quality level = MQ200
:217.09 g/mol
CAS Number:66255-85-4
Formula:C₁₀H₁₀Cl₂O