Organic reference standard, Decafluorobiphenyl 2,000 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
Catalog # EHERYA12092500MB
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:434-90-2
- Description:Decafluorobiphenyl 2,000 µg/ml in tert-butyl methyl ether
- Formula:C₁₂F₁₀
- Description:Decafluorobiphenyl